Skip to Content
Merck
All Photos(1)

Key Documents

466840

Sigma-Aldrich

Dichloromethylphosphine

96%

Synonym(s):

Methylphosphinic dichloride, Methylphosphinous dichloride, Methylphosphonous dichloride, Methylphosphorus dichloride, Methyldichlorophosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3PCl2
CAS Number:
Molecular Weight:
116.91
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

Assay

96%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.496 (lit.)

bp

82 °C (lit.)

density

1.322 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CP(Cl)Cl

InChI

1S/CH3Cl2P/c1-4(2)3/h1H3

InChI key

CDPKWOKGVUHZFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service