Skip to Content
Merck
All Photos(1)

Documents

456470

Sigma-Aldrich

Allylbromodimethylsilane

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2Si(CH3)2Br
CAS Number:
Molecular Weight:
179.13
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

95%

impurities

<3% diallyldimethylsilane

refractive index

n20/D 1.457 (lit.)

bp

130 °C (lit.)

density

1.155 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(Br)CC=C

InChI

1S/C5H11BrSi/c1-4-5-7(2,3)6/h4H,1,5H2,2-3H3

InChI key

TXEGFNIHBZYLTQ-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-9.0 °F

Flash Point(C)

-22.8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service