Skip to Content
Merck
All Photos(1)

Key Documents

451290

Sigma-Aldrich

Strontium sulfate

99.9%, powder

Synonym(s):

Celestite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrSO4
CAS Number:
Molecular Weight:
183.68
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.9%

form

powder

density

3.96 g/mL at 25 °C (lit.)

SMILES string

[Sr++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.Sr/c1-5(2,3)4;/h(H2,1,2,3,4);/q;+2/p-2

InChI key

UBXAKNTVXQMEAG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service