Skip to Content
Merck
All Photos(1)

Key Documents

430293

Sigma-Aldrich

Castor oil glycidyl ether

Synonym(s):

9-Octadecenoic acid, 12-(oxiranylmethoxy)-, 1,2,3-propanetriyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

form

viscous liquid

eq. wt.

550-650 Epoxide

refractive index

n20/D 1.479 (lit.)

viscosity

300 cP(25 °C)(lit.)

density

1.02 g/mL at 25 °C (lit.)

InChI

1S/C66H116O12/c1-4-7-10-31-40-57(70-49-60-52-73-60)43-34-25-19-13-16-22-28-37-46-64(67)76-55-63(78-66(69)48-39-30-24-18-15-21-27-36-45-59(42-33-12-9-6-3)72-51-62-54-75-62)56-77-65(68)47-38-29-23-17-14-20-26-35-44-58(41-32-11-8-5-2)71-50-61-53-74-61/h25-27,34-36,57-63H,4-24,28-33,37-56H2,1-3H3

InChI key

ZFJYZDDXGKWNCH-UHFFFAOYSA-N

Application

Shock resistant potting and dip coating compounds, high impact tooling compounds and high peel strength adhesives.

Features and Benefits

Imparts flexibility, impact resistance and shock resistance to epoxy formulations. Polymerized with amines, anhydrides, and protonic and Lewis acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service