Skip to Content
Merck
All Photos(1)

Key Documents

418277

Sigma-Aldrich

Aluminum 2,9,16,23-tetrakis(phenylthio)-29H,31H-phthalocyanine chloride

Dye content 90 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H32AlClN8S4
CAS Number:
Molecular Weight:
1007.60
UNSPSC Code:
12352103
PubChem Substance ID:

composition

Dye content, 90%

mp

210 °C (dec.) (lit.)

λmax

725 nm

SMILES string

[Cl-].[Al+]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7cc(Sc8ccccc8)ccc37)c9ccc(Sc%10ccccc%10)cc69)c%11ccc(Sc%12ccccc%12)cc5%11)c%13ccc(Sc%14ccccc%14)cc4%13

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service