Skip to Content
Merck
All Photos(1)

Key Documents

376396

Sigma-Aldrich

N,N,N′,N′-Tetramethyl-D-tartaramide

98%

Synonym(s):

(−)-N,N,N′,N′-Tetramethyl-D-tartaric acid diamide, (−)-D-Tartaric acid bis(dimethylamide), (S,S)-(−)-2,3-Dihydroxy-N,N,N′,N′-tetramethylsuccinamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH(OH)CON(CH3)2]2
CAS Number:
Molecular Weight:
204.22
Beilstein:
4675378
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D −46°, c = 3 in ethanol

mp

186-188 °C (lit.)

SMILES string

CN(C)C(=O)[C@@H](O)[C@H](O)C(=O)N(C)C

InChI

1S/C8H16N2O4/c1-9(2)7(13)5(11)6(12)8(14)10(3)4/h5-6,11-12H,1-4H3/t5-,6-/m0/s1

InChI key

PCYDYHRBODKVEL-WDSKDSINSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service