Skip to Content
Merck
All Photos(1)

Key Documents

371610

Sigma-Aldrich

3-[Bis(glycidyloxymethyl)methoxy]-1,2-propanediol

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O7
CAS Number:
Molecular Weight:
278.30
MDL number:

grade

technical grade

SMILES string

OCC(O)COC(COCC1CO1)COCC2CO2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service