Skip to Content
Merck
All Photos(1)

Key Documents

338915

Sigma-Aldrich

Morpholinosulfur trifluoride

Synonym(s):

Morph-DAST, Morpho-DAST

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C4H8F3NOS
CAS Number:
Molecular Weight:
175.17
Beilstein:
606224
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.454 (lit.)

bp

41-42 °C/0.5 mmHg (lit.)

density

1.436 g/mL at 25 °C (lit.)

SMILES string

FS(F)(F)N1CCOCC1

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

recommended

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service