Skip to Content
Merck
All Photos(1)

Key Documents

308722

Sigma-Aldrich

Benzene-D6

99.6 atom % D

Synonym(s):

Hexadeuterobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D6
CAS Number:
Molecular Weight:
84.15
Beilstein:
1905426
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.6 atom % D

Quality Level

Assay

≥99.0%

expl. lim.

0.34-6.3 % (lit.)

packaging

pk of 5 × 0.5 mL

refractive index

n20/D 1.497 (lit.)

bp

79.1 °C/760 mmHg (lit.)

mp

6.8 °C (lit.)

density

0.950 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]c1c([2H])c([2H])c([2H])c([2H])c1[2H]

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H/i1D,2D,3D,4D,5D,6D

InChI key

UHOVQNZJYSORNB-MZWXYZOWSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resonance Raman scattering of benzene and benzene-d6 with 212.8 nm excitation.
Ziegler LD and Hudson B.
J. Chem. Phys. , 74(2), 982-992 (1981)
G Wittko et al.
The Journal of chemical physics, 123(1), 014506-014506 (2005-07-23)
The Soret coefficients S(T) of the liquids acetone, benzene, benzene-d1, 1,3,5-benzene-d3, benzene-d5, benzene-13C6, benzene-d6, n-hexane, toluene, 1,2,3,4-tetrahydronaphthalene, isobutylbenzene, and 1,6-dibromohexane have been measured in protonated and perdeuterated cyclohexane by a transient holographic grating technique. It has been found that S(T)
Infrared intensities of liquids XXII: Optical and dielectric constants, molar polarizabilities, and integrated intensities of liquid benzene-d6 at 25?C between 5000 and 450 cm-1.
Bertie JE and Keefe CD.
Fresenius Journal of Analytical Chemistry, 362(1), 91-108 (1998)
The conformational distribution in diphenylmethane determined by nuclear magnetic resonance spectroscopy of a sample dissolved in a nematic liquid crystalline solvent.
Celebre G, et al.
J. Chem. Phys. , 118(14), 6417-6426 (2003)
Zachariah M Norman et al.
ACS nano, 8(7), 7513-7521 (2014-06-25)
We report the evolution of electrical transport and grain size during the sintering of thin films spin-cast from soluble phosphine and amine-bound, chloride-terminated cadmium selenide nanocrystals. Sintering of the nanocrystals occurs in three distinct stages as the annealing temperature is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service