Skip to Content
Merck
All Photos(1)

Key Documents

280569

Sigma-Aldrich

Trichlorododecylsilane

98%

Synonym(s):

Dodecyltrichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11SiCl3
CAS Number:
Molecular Weight:
303.77
Beilstein:
1758004
EC Number:
MDL number:
UNSPSC Code:
12352300

Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.454 (lit.)

bp

293-294 °C (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC[Si](Cl)(Cl)Cl

InChI

1S/C12H25Cl3Si/c1-2-3-4-5-6-7-8-9-10-11-12-16(13,14)15/h2-12H2,1H3

InChI key

BNCXNUWGWUZTCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service