Skip to Content
Merck
All Photos(1)

Documents

245216

Methyl 2-(chlorosulfonyl)benzoate

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO4S
CAS Number:
Molecular Weight:
234.66
PubChem Substance ID:

grade

technical grade

Assay

90%

InChI

1S/C8H7ClO4S/c1-13-8(10)6-4-2-3-5-7(6)14(9,11)12/h2-5H,1H3

InChI key

HUNUAFNLLYVTQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service