Skip to Content
Merck
All Photos(1)

Key Documents

177954

Sigma-Aldrich

Methyl methylthiomethyl sulfoxide

97%

Synonym(s):

Formaldehyde dimethyl mercaptal S-oxide, MMTS, Methyl methylmercaptomethyl sulfoxide, Methyl methylsulfinylmethyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SOCH2SCH3
CAS Number:
Molecular Weight:
124.23
Beilstein:
906789
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

97%

SMILES string

CSCS(C)=O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service