Skip to Content
Merck
All Photos(1)

Documents

13450

Sigma-Aldrich

Benzylhydrazine oxalate salt

≥96.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHNH2 · (COOH)2
CAS Number:
Molecular Weight:
212.20
Beilstein:
3759765
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0%

form

solid

mp

183-188 °C (dec.)

SMILES string

OC(=O)C(O)=O.NNCc1ccccc1

InChI

1S/C7H10N2.C2H2O4/c8-9-6-7-4-2-1-3-5-7;3-1(4)2(5)6/h1-5,9H,6,8H2;(H,3,4)(H,5,6)

InChI key

RCHUSXHCJLZTEY-UHFFFAOYSA-N

Application

Benzylhydrazine oxalate salt was used to study the reactions of bovine serum amine oxidase with benzylhydrazine.

Biochem/physiol Actions

Benzylhydrazine inhibits irreversibly human brain type A and type B monoamine oxidase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzylhydrazine--a selective inhibitor of human and rat brain monoamine oxidase.
J A Roth
Biochemical pharmacology, 28(6), 729-732 (1979-03-15)
L Morpurgo et al.
The Biochemical journal, 260(1), 19-25 (1989-05-15)
Bovine serum amine oxidase is inhibited by benzylhydrazine (BHy), but recovers full activity after a few hours incubation [Hucko-Haas & Reed (1970) Biochem. Biophys. Res. Commun. 38, 396-400]. The first phase of the process, requiring about 15 min, was found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service