Skip to Content
Merck
All Photos(1)

Documents

106305

Sigma-Aldrich

Nitrilotriacetic acid trisodium salt monohydrate

≥99%

Synonym(s):

Sodium nitrilotriacetate tribasic monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N(CH2COONa)3 · H2O
CAS Number:
Molecular Weight:
275.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

mp

>300 °C (lit.)

SMILES string

O.[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)CC([O-])=O

InChI

1S/C6H9NO6.3Na.H2O/c8-4(9)1-7(2-5(10)11)3-6(12)13;;;;/h1-3H2,(H,8,9)(H,10,11)(H,12,13);;;;1H2/q;3*+1;/p-3

InChI key

HERBOKBJKVUALN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Nitrilotriacetic acid trisodium salt monohydrate was used to develop capillary zone electrophoretic method for determination of eight carboxylates as copper complexes.

Biochem/physiol Actions

Nitrilotriacetic acid trisodium salt monohydrate is a potent nongenotoxic carcinogen and has induced tumors of the urinary bladder and kidney on long term administration in rats. It induces diphtheria toxin-resistant mutants in human cell lines. It inhibits gap-junctional communication between chinese hamster lung fibroblast.

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Malcolm et al.
Cell biology and toxicology, 5(2), 145-153 (1989-06-01)
Di(2-ethylhexyl)phthalate and trisodium nitrilotriacetate monohydrate, two apparently nongenotoxic carcinogens, were tested for effects on gap-junctional communication between Chinese hamster V79 lung fibroblasts. Both compounds inhibited gap-junctional communication in a concentration-dependent manner. The inhibiting effects of these chemicals on gap-junctional communication
S Inoue et al.
Cancer research, 47(24 Pt 1), 6522-6527 (1987-12-15)
Reactivities of Fe3+ chelates of aminopolycarboxylic acids with DNA were investigated by the DNA-sequencing technique using 32P 5'-end-labeled DNA fragments obtained from the human c-Ha-ras-1 protooncogene, and the reaction mechanism was studied by electron spin resonance spectroscopy. Ferric nitrilotriacetate (Fe3+-NTA)
Pirkko-Leena Laamanen et al.
Journal of chromatography. A, 1095(1-2), 164-171 (2005-11-09)
Two new methods of capillary zone electrophoresis based on aqueous phosphate running buffers with UV spectrophotometric detection were developed and optimized for the determination of eight carboxylates as copper complexes. Metalcomplexes are negatively charged, so measurements were made as anion
M P Grilli et al.
Toxicology letters, 25(2), 137-141 (1985-05-01)
The mutagenic potential of nitrilotriacetic acid trisodium salt monohydrate (NTA) was evaluated on a human cell line (EUE) using selection for diphtheria toxin (DT) resistance. Cultures were treated with different doses of the chemical (2 X 10(-2)-2 X 10(-8) M)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service