Skip to Content
Merck
All Photos(1)

Documents

AC5998047283

Accela ChemBio Inc.

6-(Trifluoromethyl)imidazo[1,2-a]pyridine-3-propanoic Acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
258.2
MDL number:

Iupac Name

3-[6-(trifluoromethyl)imidazo[1,2-a]pyridin-3-yl]propanoic acid

Inchi Code

1S/C11H9F3N2O2/c12-11(13,14)7-1-3-9-15-5-8(16(9)6-7)2-4-10(17)18/h1,3,5-6H,2,4H2,(H,17,18)

InChI key

UWCPSCLZPRVXIN-UHFFFAOYSA-N

storage temp.

+4C

Purity

95%

Country of Origin

CN

Shipping Temperature

RT

Physical Form

White to yellow solid

Product Link

http://www.accelachem.com/cn/search.html?keyword=SY056767&attname=GoodsCode

MSDS

http://pdf.accelachem.com/cn/msds/SY056767.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service