GOSU20
Sulfur in synthetic diesel
5.0000% (50′000 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material (UKAS ISO 17025 & ISO 17034 certified)
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
concentration
5.0000% (50′000 ppm)
application(s)
petroleum
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
170.6 °F
Flash Point(C)
77 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service