Z719439
Sorenson™ low binding aerosol barrier tips
MultiGuard, no graduation, maximum volume 200 μL
Synonym(s):
Aerosol Barrier Tips
All Photos(1)
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
maximum volume:
200 μL
sterility:
sterile
sterile
sterile
feature:
DNase free
RNase free
barrier
RNase free
barrier
packaging:
pack of 96 ea (Racked)
manufacturer/tradename:
Sorenson 14220T
Recommended Products
material
colorless
polypropylene
sterility
sterile
sterile
feature
DNase free
RNase free
barrier
packaging
pack of 96 ea (Racked)
manufacturer/tradename
Sorenson 14220T
maximum volume
200 μL
binding type
low binding surface
Related Categories
General description
Tips feature a new surface technology designed for quantitative assays in which sample binding can significantly alter results or for critical small volume protocols that require a high degree of accuracy. Ideal for molecular biology applications, such as PCR. Inert hydrophobic barrier prevents pipettor contamination.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)