Key Documents
20943
Repeating Dispenser
for use with Hamilton all 10 μL-2.5 mL Hamilton Syringes, for use with Hamilton PB600-1
About This Item
UNSPSC Code:
41122001
manufacturer/tradename
Hamilton 83700
packaging
1 ea of
compatibility
for use with Hamilton PB600-1
for use with Hamilton all 10 μL-2.5 mL Hamilton Syringes
General description
Accurately divides samples. Discharges 1/50th of the syringe′s capacity at each push of the button. Fits all 10 μL-2.5 mL
Hamilton syringes. Order syringe separately.
Application
Repeating Dispenser has been used for catalytic determination of enzymes and metal ions using either closed-loop or continuous-flow analyzers.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Catalytic determinations of enzymes and metal ions by sample injection in closed-loop flow systems.
Ramasamy, S. M., A. Iob, and Horacio A. Mottola.
Analytical Chemistry, 51.11, 1637-1639 (1979)