03729
Conductivity Standard
conductivity 200000 μS/cm at 25 °C
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
analytical standard
Quality Level
shelf life
limited shelf life, expiry date on the label
concentration
10-25% KCl (T)
conductivity
198000-202000 μS/cm at 25 °C
density
1.063-1.133 g/cm3 at 20 °C
application(s)
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
format
single component solution
Looking for similar products? Visit Product Comparison Guide
Application
It is a standard solution for conductivity meter calibration.
standard
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - STOT RE 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service