Skip to Content
Merck
All Photos(1)

Key Documents

HG000BX5

Millipore

Peptone Powder

Sign Into View Organizational & Contract Pricing


About This Item

packaging

pkg of 25 kg (HG000BX5.25)
poly bottle of 10 × 250 g (HG000BX5.250)

storage temp.

2-30°C

Application

Biolab Peptone is prepared by the peptic digestion of animal tissue. It contains nitrogen in a form readily available for bacterial growth requirements. It is completely soluble in water, forming a straw coloured solution and is free of fermentable carbohydrates.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service