8.14561
3-Methoxy-2-cyclohexen-1-one
for synthesis
Synonym(s):
3-Methoxy-2-cyclohexen-1-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H10O2
CAS Number:
Molecular Weight:
126.15
MDL number:
UNSPSC Code:
12352115
EC Index Number:
240-837-0
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥98.5% (GC)
bp
114 °C/19 hPa
mp
32-35 °C
storage temp.
2-30°C
SMILES string
O(C)C1=CC(=O)CCC1
InChI
1S/C7H10O2/c1-9-7-4-2-3-6(8)5-7/h5H,2-4H2,1H3
InChI key
JFTPIEHHCQPVCS-UHFFFAOYSA-N
Analysis Note
Assay (GC, area%): ≥ 98.5 % (a/a)
Melting range (lower value): ≥ 32 °C
Melting range (upper value): ≤ 35 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
Melting range (lower value): ≥ 32 °C
Melting range (upper value): ≤ 35 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service