Skip to Content
Merck
All Photos(1)

Key Documents

569399-M

Millipore

1-Stearoyl-2-arachidonoyl-sn-glycerol

Diacylglycerol derivative.

Synonym(s):

1-Stearoyl-2-arachidonoyl-sn-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H72O5
CAS Number:
Molecular Weight:
645.01
MDL number:
UNSPSC Code:
12352211

Quality Level

Assay

≥95% (TLC)

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear

solubility

DMSO: 5 mg/mL
ethanol: soluble

storage temp.

−20°C

SMILES string

O([C@H](COC(=O)CCCCCCCCCCCCCCCCC)CO)C(=O)CCC\C=C/C\C=C/C\C=C/C\C=C/CCCCC

InChI

1S/C41H72O5/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-36-41(44)46-39(37-42)38-45-40(43)35-33-31-29-27-25-23-21-18-16-14-12-10-8-6-4-2/h11,13,17,19,22,24,28,30,39,42H,3-10,12,14-16,18,20-21,23,25-27,29,31-38H2,1-2H3/b13-11-,19-17-,24-22-,30-28-/t39-/m0/s1

InChI key

NSXLMTYRMFVYNT-IUJDHQGTSA-N

General description

Diacylglycerol derivative. Endogenous activator of protein kinase C. Produced via receptor-mediated inositol phospholipid hydrolysis.

Biochem/physiol Actions

Primary Target
Protein kinase C

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Vernhet, L., et al. 1997. Life Sci. 61, 1667.
Go, M., et al. 1987. Biochem. Biophys. Res. Commun.144, 598.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service