1.18349
Calibration Standard Set
turbidity 0.02-1750 NTU
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
47101516
NACRES:
NA.24
Recommended Products
technique(s)
photometry: suitable
turbidity
0.02-1750 NTU
compatibility
for use with Turbiquant™ 3000 T
storage temp.
15-25°C
General description
0.02 - 10.0 - 100.0 - 1750 NTU, Turbiquant™ 3000 T
The Turbiquant™ Calibration Standard Set is suitable to calibrate the [Turbiquant"] 3000 T. The standards offer the following advantages:
The Turbiquant™ Calibration Standard Set is suitable to calibrate the [Turbiquant"] 3000 T. The standards offer the following advantages:
- The new primary standards have approvals which secondary standards do not have (rated to be a primary standard according to US EPA)
- Instruments used in drinking water analysis no longer need to be certified with separate primary standards
- Clear records of final inspection are included in each set
- The new standards have an unrivalled precision due to production accuracy, and stability in solution
- The standards are ready-to-use
- They are non-toxic
- They are unproblematic to store and transport
Analysis Note
Measured value (standard 1)0.02 FNU ±0.02 FNUMeasured value (standard 2)10.0 FNU ±1 %Measured value (standard 3)100 FNU ±1 %Measured value (standard 4)1750 FNU ±2 %
Legal Information
TURBIQUANT is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service