Skip to Content
Merck
All Photos(4)

Key Documents

04-790-S

Sigma-Aldrich

Anti-dimethyl-Histone H3 (Lys4) Antibody, clone AW30, Trial Size

culture supernatant, clone AW30, Upstate®

Synonym(s):

H3K4me2, Histone H3 (di methyl K4)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
AW30, monoclonal
application:
ChIP
DB
WB
inhibition assay
species reactivity:
human, chicken
technique(s):
ChIP: suitable (ChIP-seq)
dot blot: suitable
inhibition assay: suitable (peptide)
western blot: suitable
citations:

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

AW30, monoclonal

species reactivity

human, chicken

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

dimethylation (Lys4)

Gene Information

human ... H3C1(8350)

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome, the basic subunit of chromatin. Histones are modified post-translationally by the actions of enzymes in both the nucleus and cytoplasm. These modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. These modifications occur predominantly on the N-terminal and C-terminal tails that extend beyond the nucleosome core particle. Di- and trimethylation of histone H3 at Lys4 correlates with transcriptional activity of many genes. Dimethylation of Histone H3 at Lys4 occurs at both active and inactive euchromatic regions but not in silent heterochromatic sites, whereas trimethylation at Lys4 is present exclusively at active genes.

Specificity

Histone H3 dimethylated on lysine 4
The immunizing sequence is conserved from Tetrahymena to human, so broad species cross-reactivity is expected.

Immunogen

KLH-conjugated, synthetic peptide containing the sequence …Tme2KQT… in which me2K corresponds to dimethyl-lysine at residue 4 of human histone H3.

Application

Western Blot Analysis:
A 1:1000-1:2000 dilution of this lot detected dimethyl histone H3 (Lys4) in HeLa acid extracts.

Western Blot Analysis:
A 1:2,000 to 1:10,000 dilution of a previous lot detected methylated histone H3 in acid extracted proteins from HeLa cells. The antibody did not detect unmethylated recombinant Histone H3.

ChIP-seq Analysis:
Representative lot data. Chromatin immunoprecipitation was performed using the Magna ChIP HiSens kit (cat# 17-10460), Anti-dimethyl-Histone H3 (Lys4) antibody (2 µl cat# 04-790-S), 20 µL Protein A/G beads, and 1e6 crosslinked HeLa cell chromatin followed by DNA purification using magnetic beads. Libraries were prepared from Input and ChIP DNA samples using standard protocols with Illumina barcoded adapters, and analyzed on Illumina HiSeq instrument. An excess of sixteen million reads from FastQ files were mapped using Bowtie (http://bowtie-bio.sourceforge.net/manual.shtml) following TagDust (http://genome.gsc.riken.jp/osc/english/dataresource/) tag removal. Peaks were identified using MACS (http://luelab.dfci.harvard.edu/MACS/), with peaks and reads visualized as a custom track in UCSC Genome Browser (http://genome.ucsc.edu) from BigWig and BED files. The highest 25% of peaks identified in the 04-790 and 05-1338 datasets showed 92 and 90% overlap with peaks identified in the ENCODE H3K4me2 BROAD Histone track for HeLa S3.

Dot Blot Analysis:
Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665), which contain histone peptides with various modifications were probed with Cat. No 04-790, Anti-dimethyl Histone H3 (Lys4), clone AW30 at 1:1000 dilution. Proteins were visualized using a Donkey anti-Rabbit IgG conjugated to HRP and a chemiluminescence detection system.

Peptide Inhibition Analysis:
2 μM of a histone H3 peptide containing dimethyl-lysine 4 abolished detection of histone H3 by a previous lot of antibody in immunoblot analysis of HeLa acid extracts. No signal reduction was observed with histone H3 peptides containing either monomethyl or trimethyl-lysine 4 modifications
Anti-dimethyl-Histone H3 (Lys4) Antibody, clone AW30 is a rabbit monoclonal antibody for detection of dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4) & has been validated in WB, ChIP, ChIP-seq, DB, PIA, Mplex.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

routinely evaluated by immunoblot on acid-extracted proteins from HeLa cells; the antibody did not detect unmethylated recombinant Histone H3 (Catalog #14-494)

Linkage

Replaces: 05-790

Physical form

Cultured supernantant in 0.05% sodium azide

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
For maximum recovery of product, centrifuge the vial prior to removing the cap.

Analysis Note

Control
HeLa acid extracts

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service