Skip to Content
Merck
All Photos(1)

Key Documents

Z308544

Quickfit® sintered bottle heads

for use with 250 mL bottles

Synonym(s):

MF48/23/250

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51

manufacturer/tradename

Quickfit MF 48/23/250

availability

not available in USA and Canada

compatibility

for use with 250 mL bottles

Looking for similar products? Visit Product Comparison Guide

General description

Sintered heads improve gas dispersion

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service