SYNF311910
Synthware™ recovery flask
10 mL, joint: ST/NS 19/22
Synonym(s):
Synthware F311910, oval shape flask
All Photos(1)
About This Item
material
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F311910
capacity
10 mL
10 mL
Related Categories
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.