Skip to Content
Merck
All Photos(1)

Key Documents

SLW3722/08M

Pyrex® Büchner filter funnels with sintered disc

4000 mL, disc diam. 120 mm, porosity grade 4

Synonym(s):

Pyrex, SciLabware 3722/08M

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.43

manufacturer/tradename

3722/08M
SciLabware

capacity

4000 mL
4000 mL

disc diam.

120 mm

pore size

grade 4 porosity

Looking for similar products? Visit Product Comparison Guide

General description

The 175mm sintered discs fitted to these funnels are manufactured from the same 3.3 expansion borosilicate glass as the funnel bodies, making these funnels suitable to be used with a wide range aggressive chemicals.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service