SLW3151/26
Pyrex® Volumetric flask, heavy duty, class A, USP/ISO/DIN tolerances
capacity 5000 mL
Synonym(s):
Pyrex, SciLabware 3151/26
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
Pyrex flask
Joint
joint ST/NS neck34/35
feature
accuracy class A (ISO 1042 and DIN 12664, United States Pharmacopoeia (USP),conformity ′H′ certified)
packaging
pkg of 1 ea
manufacturer/tradename
SciLabware 3151/26
Overall H
475 mm
capacity
5000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Class A, Pyrex
- Complies with ISO 1042 and DIN 12664
- Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which is tighter than those of ISO/DIN standards
- Each flask is laser etched with an individual serial number, batch code and date of manufacture
- Conformity ′H′, certified
- Manufactured from chemically resistant Pyrex glass
- Inscriptions in white enamel
- Polyethylene stopper
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service