Skip to Content
Merck
All Photos(2)

Documents

GF22210108

Nickel

foil, thickness 0.125 mm, size 150 × 150 mm, purity 99%

Synonym(s):

NI000501

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

foil

manufacturer/tradename

Goodfellow 222-101-08

resistivity

6.97 μΩ-cm, 20°C

size

150 × 150 mm

thickness

0.125 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service