Skip to Content
Merck
All Photos(1)

Documents

59202

Supelco

Postcolumn Reactor Components, Glass Beads

size 250 μm, acid-washed

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

packaging

pkg of 25 g

size

250 μm

Looking for similar products? Visit Product Comparison Guide

General description

The equipment needed to perform post-column reactions can be relatively simple. These components (glass beads, acid-washed) enable easy and economic construction of desired systems. For large peak volumes, the use of a column (5 cm x 4.6 mm) filled with 250 μm beads is recommended. For small peak volumes, a single bead string reactor (30 cm of 0.5 mm I.D. PTFE tubing) filled with 250 μm beads may be utilized.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service