SAB4701098
Monoclonal Anti-CD3-APC antibody produced in mouse
clone APA1/1
Synonym(s):
CD3 epsilon chain, CD3epsilon, IMD18, T3E
About This Item
Recommended Products
biological source
mouse
conjugate
Allophycocyanin conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
APA1/1, monoclonal
form
buffered aqueous solution
concentration
0.1 mg/mL
technique(s)
flow cytometry: 1.5 μg/mL
accession no.
BC049847
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... CD3E(916)
Related Categories
General description
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service