MAK491
Formate Assay Kit
sufficient for 100 colorimetric tests
Synonym(s):
Formate Test Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
Quality Level
application(s)
environmental
detection method
colorimetric
relevant disease(s)
neurological disorders
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
Formate is the anion derived from formic acid, the simplest carboxylic acid. It is also the metabolic byproduct of formaldehyde metabolism in the body, and the eventual metabolic byproduct of methanol, which is first broken down to formaldehyde. At high levels, formate is neurotoxic to the central nervous system and can cause blindness, coma, and death. Although naturally present in the body at low levels, elevated levels of formate may be used as an indicator of formaldehyde exposure and methanol poisoning.
Features and Benefits
- Linear detection range 0.050 to 5 mM formate.
- Compatible with high-throughput handling systems.
Suitability
The kit is suitable for the determination of formate in biological samples such as urine and serum.
Principle
The Formate Assay Kit is based on the formate dehydrogenase catalyzed oxidation of formate, which generates carbon dioxide and NADH that reduces a formazan (MTT) dye. The intensity of the reduced MTT, measured at 565 nm, is directly proportional to the formate concentration in the sample.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service