Skip to Content
Merck
All Photos(1)

Key Documents

CLS3821BC

Corning® 384 well microplate, low volume

black polystyrene, flat bottom, non-sterile, with generic barcode, lid: no, pack of 10

Synonym(s):

384 multiwell plate, 384 well barcoded plate, 384 well microplate, 384 well microtiter plate, 384 well plate, 384 well plate with generic barcode

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
black polystyrene
flat bottom
flat bottom black polystyrene wells
well volume:
50 μL
wells:
384 wells , low volume
working volume:
5-40 μL
Pricing and availability is not currently available.

material

black polystyrene
flat bottom
flat bottom black polystyrene wells

sterility

non-sterile

feature

lid: no
skirt
with generic barcode

packaging

pack of 10

manufacturer/tradename

Corning 3821BC

well volume

50 μL

wells

384 wells , low volume

working volume

5-40 μL

suitability

suitable for (molecular, validated for the FLIPR reader)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Microplate Features
  • Round well design with a solid black flat bottom
  • 2.8 mm distance between well bottoms to plate bottom
  • Validated by Molecular Devices for the FLIPR reader. Outer wells aligned for proper viewing with FLIPR
  • Ideal for miniaturized 384 well fluorescent homogeneous assays
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200 ng IgG/cm2)

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service