Skip to Content
Merck
All Photos(1)

Documents

CRFK (BVD Ag negative)

86093002, feline kidney, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CRFK (BVD Ag negative), 86093002

biological source

feline kidney

growth mode

Adherent

karyotype

Hypodiploid, modal no 36

morphology

Epithelial

products

Carcinoembryonic antigen (CEA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Cat kidney

Cell Line Description

CRFK cells were isolated from the cortical portion of the kidneys of a 10-12 week old normal female domestic cat. CRFK are extensively used in feline virus research. Susceptible to herpesvirus and feline picornaviruses producing CPE. BVD Ag negative.

Application

Crandell feline kidney cells (CRFK) (BVD Ag negative) have been used in viral studies.

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:8 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service