Skip to Content
Merck
All Photos(1)

Key Documents

NS6-15-05PK100

Millipore

NovaSeptum® Sealing

NovaSeptum® Pinch Pipes for 3-6mm (ID-OD) Tubes - 100 pack

Synonym(s):

NovaSeptum® Tube Disconnection Pinch Pipes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
eCl@ss:
32010310

sterility

non-sterile

product line

NovaSeptum®

application(s)

sterile sampling

compatibility

for use with NovaSeptum® crimping tool. A100-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service