MABC1099
Anti-uPAR (CD87) Antibody, clone 5D6
clone 5D6, from mouse
Synonym(s):
Urokinase plasminogen activator surface receptor, CD87
About This Item
Recommended Products
biological source
mouse
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
5D6, monoclonal
species reactivity
hamster
packaging
antibody small pack of 25 μg
technique(s)
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
hamster ... Plaur(100689422)
General description
Specificity
Immunogen
Application
Apoptosis & Cancer
Flow Cytometry Analysis: A representative lot detected uPAR (CD87) in Flow Cytometry applications (Ashida, H., et. al. (2006). J Biol Chem. 281(2):896-904; Lee, G.H., et. al. (2016). J Cell Biol. 215(5):705-718; Hirata, T., et. al. (2018). Nat Commun. 9(1):405; Hirata, T., et. al. (2013). J Biochem. 154(3):257-64).
Immunocytochemistry Analysis: A representative lot detected uPAR (CD87) in Immunocytochemistry applications (Kanzawa, N., et. al. (2012). J Lipid Res. 53(4):653-63).
Quality
Flow Cytometry Analysis: 1 µg of this antibody detected uPAR (CD87) in one million Chinese hamster ovary (CHO) cells.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service