Skip to Content
Merck
All Photos(1)

Key Documents

8.18473

Sigma-Aldrich

N,N-Diethylhydroxylamine

for synthesis

Synonym(s):

N,N-Diethylhydroxylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NOH
CAS Number:
Molecular Weight:
89.14
MDL number:
UNSPSC Code:
12352116
EC Index Number:
223-055-4

vapor pressure

33 hPa ( 20 °C)

Assay

≥98.0% (GC)

form

liquid

potency

2190 mg/kg LD50, oral (Rat)
1300 mg/kg LD50, skin (Rabbit)

expl. lim.

1.9-10 % (v/v)

reaction suitability

reagent type: reductant

bp

125-130 °C/1013 hPa

mp

-25 °C

transition temp

flash point 45 °C

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H11NO/c1-3-5(6)4-2/h6H,3-4H2,1-2H3

InChI key

FVCOIAYSJZGECG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.868 - 0.872
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(C)

45 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service