8.14986
DL-Leucine
for synthesis
Synonym(s):
DL-Leucine, DL-2-Aminmo-4-methylvalerianic acid, DL-2-Aminmo-4-methylpentanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H13NO2
CAS Number:
Molecular Weight:
131.17
MDL number:
UNSPSC Code:
12352209
EC Index Number:
206-328-2
NACRES:
NA.22
Recommended Products
Analysis Note
Assay (HClO₄): ≥ 98.0 %
Sulfated ash: ≤ 0.10 %
Identity (IR): passes test
Sulfated ash: ≤ 0.10 %
Identity (IR): passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service