66612-M
Urea
End product of the urea cycle. A topical keratolytic agent.
Synonym(s):
Carbimide, Carbonyl Diamide, Carbamide, Carbonyldiamide
About This Item
Quality Level
Assay
≥98%
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated
impurities
≤0.01% insoluble material
color
white
mp
132-135 °C (lit.)
solubility
water: soluble
density
1.335 g/mL at 25 °C (lit.)
cation traces
heavy metals: ≤0.001%
storage temp.
10-30°C
SMILES string
NC(N)=O
InChI
1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)
InChI key
XSQUKJJJFZCRTK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Warning
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service