Key Documents
Z742595
Accuris SmartDoc narrow band pass filter
535nm filter; Used for imaging green stains
Synonym(s):
Accuris SmartDoc
About This Item
UNSPSC Code:
41106000
NACRES:
NB.24
feature
535nm filter; Used for imaging green stains
packaging
pkg of 1
manufacturer/tradename
Benchmark Scientific E5001-535
General description
A narrow band pass filter of 535 nm used for imaging green stains on a UV transilluminator.
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
SmartDoc is a trademark of Benchmark Scientific, Inc.