Skip to Content
Merck
All Photos(1)

Key Documents

Z282774

Sigma-Aldrich

Lab spoons

nickel-stainless steel, L 9 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122403
NACRES:
NB.35

material

nickel-stainless steel

packaging

pkg of 6 ea

L

9 in.

spoon end size

1/2 in. × 1 in.

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

One spoon end, and one blade (5/16 × 2in.)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service