Skip to Content
Merck
All Photos(1)

Key Documents

BR781342

BRAND® strips of 8 caps

polypropylene, yellow, for PCR tubes

Synonym(s):

PCR, PCR Tubes, Tube Strips, Tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
Pricing and availability is not currently available.

material

domed
polypropylene
yellow

feature

for PCR tubes

packaging

pack of 125 ea
pack of 125 ea

manufacturer/tradename

BRAND 781342

technique(s)

PCR: suitable

Looking for similar products? Visit Product Comparison Guide

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service