Skip to Content
Merck
All Photos(1)

Documents

SAAR5945020

L-(+)-Tartaric acid

univAR

Synonym(s):

(2R,3R)-(+)-Tartaric acid, L-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725147
MDL number:
UNSPSC Code:
12352106

vapor density

5.18 (vs air)

product line

univAR

autoignition temp.

797 °F

packaging

poly bottle of 12 × 500 g (SAAR5945020EM)

mp

170-172 °C (lit.)

storage temp.

2-30°C

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1

InChI key

FEWJPZIEWOKRBE-JCYAYHJZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service