Skip to Content
Merck
All Photos(1)

Key Documents

SAAR2862000

Heptane

uniLAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1730763
MDL number:
UNSPSC Code:
12352001

vapor density

3.5 (vs air)

vapor pressure

40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

product line

uniLAB

autoignition temp.

433 °F

expl. lim.

7 %

packaging

glass bottle of 4 × 2.5 L (SAAR2862000LC)

refractive index

n20/D 1.387 (lit.)

bp

98 °C (lit.)

mp

−91 °C (lit.)

density

0.684 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCCCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chemically pure
For laboratory use only

Legal Information

uniLAB is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service