SAAR190696
Hydrochloric acid solution
c(HCL) = 0.5 mol/l (0.5 N)
Synonym(s):
Hydrochloric acid solution, Hydrogen chloride solution
About This Item
Recommended Products
Product Name
Hydrochloric acid solution 0.5N (0.5M),
packaging
poly bottle of 1 × 5 L (SAAR190696LL)
density
1.01 g/mL
storage temp.
2-30°C
SMILES string
Cl
InChI
1S/ClH/h1H
InChI key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
nwg
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service