908894
Nishimura′s catalyst
Umicore
Synonym(s):
Rhodium(III) oxide/Platinum(IV) oxide hydrate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
O3Rh2/O2Pt
UNSPSC Code:
12171602
NACRES:
NA.22
Recommended Products
form
powder
Related Categories
Application
Nishimura′s catalyst is a homogeneous catalyst useful for hydrogenation reactions.
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Ox. Sol. 1
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service