Skip to Content
Merck
All Photos(1)

Key Documents

AB5883

Sigma-Aldrich

Anti-Synapsin I Antibody, phosphoSer 603

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
rat
technique(s):
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer603)

Gene Information

human ... SYN1(6853)

Specificity

Synapsin I, phosphoSer603. This is the site phosphorylated by Calcium calmodulin kinase II. The antibody recognizes the ~78 kD Synapsin I, phosphoSer603 protein. Immunolabeling is blocked by the immunogen peptide but no by the corresponding non-phospho peptide.

Immunogen

Epitope: phosphoSer 603
Synthetic peptide corresponding to amino acids surrounding the phosphoSer603 of rat Synapsin I. The immunogen sequence is identical in human, mouse and Xenopus.

Application

Anti-Synapsin I Antibody, phosphoSer 603 detects level of Synapsin I & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Western blot: 1:1,000. Rat brain lysate shows a ~78-80kDa band.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin presented as a liquid in 10 mM HEPES, pH 7.5, with 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.

Storage and Stability

Maintain at -20°C in undiluted for up to 6 months from date of receipt. Avoid repeated freeze/thaw cycles. Do not store in a self-defrosting freezer.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie Fourneau et al.
Journal of neurochemistry, 147(2), 240-255 (2018-05-29)
In human, a chronic sensorimotor perturbation (SMP) through prolonged body immobilization alters motor task performance through a combination of peripheral and central factors. Studies performed on a rat model of SMP have shown biomolecular changes and a reorganization of sensorimotor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service