Z231711
Duran® crystallizing dishes
capacity 150 mL
Synonym(s):
213134104
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
borosilicate glass 3.3
packaging
pack of 1 ea
case of 10 ea
manufacturer/tradename
DURAN® 213134104
availability
not available in USA and Canada
capacity
150 mL
dish diam.
80 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Cylindrical dishes without spout.
- Manufactured from Duran borosilicate glass type 3.3 for maximum chemical and thermal shock resistance.
- The design of the dishes allow solvents to evaporate quickly and evenly
- Fired-on, highly durable white ceramic
- Meets DIN 12337 standard.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service