DWKWGW536PML
Azlon Multi-Lingual Unvented Wash Bottles
label, Water (in English, French, Spanish), capacity 500 mL
Synonym(s):
WGW536PML
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103209
NACRES:
NB.43
Recommended Products
feature
label Water (in English, French, Spanish)
packaging
pkg of 5 ea
manufacturer/tradename
Azlon WGW536PML
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ideal for water dispensing within the laboratory
Features and Benefits
Multilingual with solvent labels including all new globally harmonised symbols, NFPA diamond, risk phrases and safety information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service