BAF369740950
SP Bel-Art® Sterileware™ Powder Lance Samplers
white high-density polyethylene, capacity 200 mL (6.8 oz), L 95 cm (37.5 in.), sterile, pack of 20 ea (individually wrapped)
Synonym(s):
sampler
All Photos(4)
About This Item
Recommended Products
material
white high-density polyethylene
sterility
sterile
packaging
pack of 20 ea (individually wrapped)
manufacturer/tradename
Bel-Art F36974-0950
L
95 cm (37.5 in.)
capacity
200 mL (6.8 oz)
General description
A simple insert, rotate, and withdraw motion allows cross sectional sampling of non-flowing, bulk powders. Available in three lengths.
Features and Benefits
- Open blade works well with sticky, cohesive powders
- Rounded edges help hold sample in place
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) lance is heat sealed in individual polyethylene bag
- Gamma irradiated sterile (SAL 10^-6)
- Ready for one time use and disposal
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)